missing translation for 'onlineSavingsMsg'
Learn More

Dimethylthiocarbamoyl chloride, 99%

Catalog No. AC408610250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC408610250 25 g Glass bottle
AC408611000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC408610250 Supplier Thermo Scientific Chemicals Supplier No. 408610250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16420-13-6 | C3H6ClNS | 123.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16420-13-6 , 79-44-7
Molecular Formula C3H6ClNS
Molecular Weight (g/mol) 123.60
MDL Number MFCD00004919
InChI Key PHWISQNXPLXQRU-UHFFFAOYSA-N
Synonym dimethylthiocarbamoyl chloride, carbamothioic chloride, dimethyl, chloromethanethioyl dimethylamine, n,n-dimethylthiocarbamoyl chloride, n,n-dimethylcarbamothioic chloride, dimethylthiocarbamoylchloride, carbamoyl chloride, dimethylthio, carbamothioic chloride, n,n-dimethyl, dimethylthiocarbamyl chloride, dimethylthiocarbamic acid chloride
PubChem CID 27871
IUPAC Name N,N-dimethylcarbamothioyl chloride
SMILES CN(C)C(Cl)=S

Specifications

Melting Point 42°C to 43°C
Color Yellow
Boiling Point 63°C to 65°C (0.2 mbar)
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCSCl
Quantity 25 g
Beilstein 04, 75
Fieser 02,173; 03,127; 04,202; 08,200
Solubility Information Solubility in water: reacts
Formula Weight 123.61
Percent Purity 99%
Physical Form Low Melting Crystalline Solid or Crystals
Chemical Name or Material Dimethylthiocarbamoyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause cancer.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 240-468-5
TSCA TSCA

RUO – Research Use Only