missing translation for 'onlineSavingsMsg'
Learn More

Dimethylthiocarbamoyl chloride, 95%

Catalog No. aah6010422
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6010422 100 g
AAH6010414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6010422 Supplier Thermo Scientific Chemicals Supplier No. H6010422

CAS: 16420-13-6 | C3H6ClNS | 123.60 g/mol

Dimethylthiocarbamoyl chloride is used as an intermediate in the synthesis of pharmaceuticals. It finds application in chemoselective deoxygenation of pyridine-N-oxides. It is also used as a starting reagent in the synthesis of dimethylthiocarbamates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylthiocarbamoyl chloride is used as an intermediate in the synthesis of pharmaceuticals. It finds application in chemoselective deoxygenation of pyridine-N-oxides. It is also used as a starting reagent in the synthesis of dimethylthiocarbamates.

Solubility
Soluble in chloroform and tetrahydrofuran.

Notes
Store in cool place. Moisture sensitive. Incompatible with strong oxidizing agents, amines and strong bases. Reacts violently with water.

Chemical Identifiers

CAS 16420-13-6
Molecular Formula C3H6ClNS
Molecular Weight (g/mol) 123.60
MDL Number MFCD00004919
InChI Key PHWISQNXPLXQRU-UHFFFAOYSA-N
Synonym dimethylthiocarbamoyl chloride, carbamothioic chloride, dimethyl, chloromethanethioyl dimethylamine, n,n-dimethylthiocarbamoyl chloride, n,n-dimethylcarbamothioic chloride, dimethylthiocarbamoylchloride, carbamoyl chloride, dimethylthio, carbamothioic chloride, n,n-dimethyl, dimethylthiocarbamyl chloride, dimethylthiocarbamic acid chloride
PubChem CID 27871
SMILES CN(C)C(Cl)=S

Specifications

Melting Point 39°C to 43°C
Boiling Point 90°C to 95°C
Flash Point 98°C (208°F)
Linear Formula (CH3)2NCSCl
Quantity 100 g
UN Number UN3261
Beilstein 635823
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform and tetrahydrofuran.
IUPAC Name N,N-dimethylcarbamothioyl chloride
Formula Weight 123.6
Percent Purity 95%
Chemical Name or Material Dimethylthiocarbamoyl chloride
Show More Show Less
Hazard Category H314-H317-H350
Hazard Statement GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P264b-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only