missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsulfonazo III, TCI America™

Catalog No. A50081G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50081G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A50081G Supplier TCI America Supplier No. A50081G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

[Spectrophotometric reagent for alkaline earth metals and indicator for the precipitation titration of SO4 with Ba]

Chemical Identifiers

CAS 14979-11-4
Molecular Formula C24H20N4O14S4
Molecular Weight (g/mol) 716.678
MDL Number MFCD00151118
InChI Key CAWGMVYQLWPWHZ-GPAWKIAZSA-N
Synonym 2,7-Bis[(4-methyl-2-sulfophenyl)azo]-1,8-dihydroxynaphthalene-3,6-disulfonic Acid, 2,7-Bis(2-sulfo-p-tolylazo)-1,8-dihydroxy-3,6-naphthalenedisulfonic Acid
PubChem CID 5743575
IUPAC Name (3Z,6Z)-3,6-bis[(4-methyl-2-sulfophenyl)hydrazinylidene]-4,5-dioxonaphthalene-2,7-disulfonic acid
SMILES CC1=CC(=C(C=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=C(C=C(C=C4)C)S(=O)(=O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O

Specifications

Quantity 1 g
Formula Weight 716.68
Physical Form Crystalline Powder
Chemical Name or Material Dimethylsulfonazo III [Spectrophotometric reagent for alkaline earth metals and indicator for the pr