missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsuberimidate dihydrochloride

Catalog No. p-7058773
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
2 g
Catalog No. Quantity
AA4324404 2 g
AA4324409 10 g
2 options

Catalog No. AA4324404

Supplier: Thermo Scientific Chemicals 04324404

Only null left

Crosslinking of proteins and poly(allylamine) | CAS: 34490-86-3 | C10H22Cl2N2O2 | 273.198 g/mol

Dimethylsuberimidate dihydrochloride is used as a homobifunctional cross-linking reagent for amino groups. It is also used for the modification of tyrosinase to produce biocatalytic enzymatic flow-through detectors. It serves as a cross-linker for preparation of collagen-calcium phosphate composites. Further, it is used in the treatment of avian myeloblastosis virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34490-86-3
Molecular Formula C10H22Cl2N2O2
Molecular Weight (g/mol) 273.198
MDL Number MFCD00012574
InChI Key ILKCDNKCNSNFMP-UHFFFAOYSA-N
Synonym dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride
PubChem CID 118696
IUPAC Name dimethyl octanediimidate;dihydrochloride
SMILES COC(=N)CCCCCCC(=N)OC.Cl.Cl

Specifications

Melting Point 213°C to 214°C
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 273.2
Physical Form Crystalline
Chemical Name or Material Dimethylsuberimidate dihydrochloride
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 252-060-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only