missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylmalonic Acid 98.0+%, TCI America™
$44.73 - $119.30
Chemical Identifiers
| CAS | 595-46-0 |
|---|---|
| Molecular Formula | C5H8O4 |
| Molecular Weight (g/mol) | 132.12 |
| MDL Number | MFCD00004193 |
| InChI Key | OREAFAJWWJHCOT-UHFFFAOYSA-N |
| Synonym | dimethylmalonic acid, 2,2-dimethylmalonic acid, propanedioic acid, dimethyl, dimethylpropanedioic acid, dimethyl-propanedioic acid, 2,2-propanedicarboxylic acid, unii-55981wkx6b, dimethyl-malonic acid, propanedioic acid dimethyl, 2,2-dimethyl-propanedioic acid |
| PubChem CID | 11686 |
| IUPAC Name | dimethylpropanedioic acid |
| SMILES | CC(C)(C(O)=O)C(O)=O |
Chemical Identifiers
| 595-46-0 | |
| 132.12 | |
| OREAFAJWWJHCOT-UHFFFAOYSA-N | |
| 11686 | |
| CC(C)(C(O)=O)C(O)=O |
| C5H8O4 | |
| MFCD00004193 | |
| dimethylmalonic acid, 2,2-dimethylmalonic acid, propanedioic acid, dimethyl, dimethylpropanedioic acid, dimethyl-propanedioic acid, 2,2-propanedicarboxylic acid, unii-55981wkx6b, dimethyl-malonic acid, propanedioic acid dimethyl, 2,2-dimethyl-propanedioic acid | |
| dimethylpropanedioic acid |
Specifications
| 595-46-0 | |
| White | |
| MFCD00004193 | |
| dimethylmalonic acid, 2,2-dimethylmalonic acid, propanedioic acid, dimethyl, dimethylpropanedioic acid, dimethyl-propanedioic acid, 2,2-propanedicarboxylic acid, unii-55981wkx6b, dimethyl-malonic acid, propanedioic acid dimethyl, 2,2-dimethyl-propanedioic acid | |
| CC(C)(C(O)=O)C(O)=O | |
| 132.12 | |
| 132.12 | |
| Crystalline Powder |
| 191°C | |
| C5H8O4 | |
| 5 g | |
| OREAFAJWWJHCOT-UHFFFAOYSA-N | |
| dimethylpropanedioic acid | |
| 11686 | |
| ≥98.0% (GC,T) | |
| Dimethylmalonic Acid |
Safety and Handling
TSCA : Yes