accessibility menu, dialog, popup

UserName

CAS RN 595-46-0

H3CCH3HOOOHO

CAS RN 595-46-0

IUPAC Name: dimethylpropanedioic acid
Synonyms: dimethylmalonic acid propanedioic acid dimethyl
Molecular Weight (g/mol): 132.12
Molecular Formula: C5H8O4
InChi Key: OREAFAJWWJHCOT-UHFFFAOYSA-N
SMILES: CC(C)(C(O)=O)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)