missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylketene Methyl Trimethylsilyl Acetal 95.0+%, TCI America™
Supplier: TCI America D178925ML
Specifications
| Dimethylketene Methyl Trimethylsilyl Acetal | |
| Colorless | |
| C8H18O2Si | |
| 25 mL | |
| 1-methoxy-2-methyl-1-trimethylsiloxy propene, methyl trimethylsilyl dimethylketene acetal, dimethylketene methyl trimethylsilyl acetal, 1-methoxy-2-methylprop-1-en-1-yl oxy trimethylsilane, silane, 1-methoxy-2-methyl-1-propenyl oxy trimethyl, 1-methoxy-1-trimethylsiloxy-2-methyl-1-propene, dimethylketene methyltrimethylsilylacetal, 1-methoxy-1-trimethylsilyloxy-2-methylpropene, 1-methoxy-2-methyl-1-propenyloxy trimethylsilane, 1-methoxy-2-methylprop-1-enoxy-trimethylsilane | |
| COC(O[Si](C)(C)C)=C(C)C | |
| 174.32 | |
| 174.32 | |
| Liquid |
| 31469-15-5 | |
| 52°C | |
| MFCD00010232 | |
| 1993 | |
| JNOGVQJEBGEKMG-UHFFFAOYSA-N | |
| [(1-methoxy-2-methylprop-1-en-1-yl)oxy]trimethylsilane | |
| 552336 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 31469-15-5 | |
| 174.32 | |
| JNOGVQJEBGEKMG-UHFFFAOYSA-N | |
| 552336 | |
| COC(O[Si](C)(C)C)=C(C)C |
| C8H18O2Si | |
| MFCD00010232 | |
| 1-methoxy-2-methyl-1-trimethylsiloxy propene, methyl trimethylsilyl dimethylketene acetal, dimethylketene methyl trimethylsilyl acetal, 1-methoxy-2-methylprop-1-en-1-yl oxy trimethylsilane, silane, 1-methoxy-2-methyl-1-propenyl oxy trimethyl, 1-methoxy-1-trimethylsiloxy-2-methyl-1-propene, dimethylketene methyltrimethylsilylacetal, 1-methoxy-1-trimethylsilyloxy-2-methylpropene, 1-methoxy-2-methyl-1-propenyloxy trimethylsilane, 1-methoxy-2-methylprop-1-enoxy-trimethylsilane | |
| [(1-methoxy-2-methylprop-1-en-1-yl)oxy]trimethylsilane |
Safety and Handling
TSCA : No