missing translation for 'onlineSavingsMsg'
Learn More

Dimethylketene Methyl Trimethylsilyl Acetal 95.0+%, TCI America™

Catalog No. p-7126929
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17895ML 5 mL
D178925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17895ML Supplier TCI America Supplier No. D17895ML
Only null left

Chemical Identifiers

CAS 31469-15-5
Molecular Formula C8H18O2Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD00010232
InChI Key JNOGVQJEBGEKMG-UHFFFAOYSA-N
Synonym 1-methoxy-2-methyl-1-trimethylsiloxy propene, methyl trimethylsilyl dimethylketene acetal, dimethylketene methyl trimethylsilyl acetal, 1-methoxy-2-methylprop-1-en-1-yl oxy trimethylsilane, silane, 1-methoxy-2-methyl-1-propenyl oxy trimethyl, 1-methoxy-1-trimethylsiloxy-2-methyl-1-propene, dimethylketene methyltrimethylsilylacetal, 1-methoxy-1-trimethylsilyloxy-2-methylpropene, 1-methoxy-2-methyl-1-propenyloxy trimethylsilane, 1-methoxy-2-methylprop-1-enoxy-trimethylsilane
PubChem CID 552336
IUPAC Name [(1-methoxy-2-methylprop-1-en-1-yl)oxy]trimethylsilane
SMILES COC(O[Si](C)(C)C)=C(C)C

Specifications

Color Colorless
Boiling Point 52°C
Quantity 5 mL
UN Number 1993
Formula Weight 174.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethylketene Methyl Trimethylsilyl Acetal