missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyldipalmitylammonium Bromide 97.0+%, TCI America™
$251.65 - $1813.73
Chemical Identifiers
| CAS | 70755-47-4 |
|---|---|
| Molecular Formula | C34H72BrN |
| Molecular Weight (g/mol) | 574.86 |
| MDL Number | MFCD00191428 |
| InChI Key | VIXPKJNAOIWFMW-UHFFFAOYSA-M |
| Synonym | Dihexadecyldimethylammonium Bromide |
| PubChem CID | 9808235 |
| IUPAC Name | dihexadecyldimethylazanium bromide |
| SMILES | [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC |
Chemical Identifiers
| 70755-47-4 | |
| 574.86 | |
| VIXPKJNAOIWFMW-UHFFFAOYSA-M | |
| 9808235 | |
| [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC |
| C34H72BrN | |
| MFCD00191428 | |
| Dihexadecyldimethylammonium Bromide | |
| dihexadecyldimethylazanium bromide |
Specifications
| 70755-47-4 | |
| C34H72BrN | |
| 25 g | |
| VIXPKJNAOIWFMW-UHFFFAOYSA-M | |
| dihexadecyldimethylazanium bromide | |
| 9808235 | |
| ≥97.0% (T) | |
| Dimethyldipalmitylammonium Bromide |
| White | |
| MFCD00191428 | |
| Dihexadecyldimethylammonium Bromide | |
| [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC | |
| 574.86 | |
| 574.86 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
TSCA : No