missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldipalmitylammonium Bromide 97.0+%, TCI America™

Catalog No. D235425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D235425G 25 g
D2354500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D235425G Supplier TCI America Supplier No. D235425G
Only null left

Chemical Identifiers

CAS 70755-47-4
Molecular Formula C34H72BrN
Molecular Weight (g/mol) 574.86
MDL Number MFCD00191428
InChI Key VIXPKJNAOIWFMW-UHFFFAOYSA-M
Synonym Dihexadecyldimethylammonium Bromide
PubChem CID 9808235
IUPAC Name dihexadecyldimethylazanium bromide
SMILES [Br-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC

Specifications

Color White
Quantity 25 g
Formula Weight 574.86
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyldipalmitylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA No