missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldioctylammonium Bromide 97.0+%, TCI America™

Supplier:  TCI America D275425G

 View more versions of this product

Catalog No. D275425G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dimethyldioctylammonium Bromide
White-Yellow
MFCD00082931
APTVNWGLSRAOFJ-UHFFFAOYSA-M
dimethyl(dioctyl)azanium;bromide
76408
≥97.0% (T)
3026-69-5
C18H40BrN
25 g
CCCCCCCC[N+](C)(C)CCCCCCCC.[Br-]
350.429
350.43
Crystalline Powder

Chemical Identifiers

3026-69-5
350.429
APTVNWGLSRAOFJ-UHFFFAOYSA-M
dimethyl(dioctyl)azanium;bromide
C18H40BrN
MFCD00082931
76408
CCCCCCCC[N+](C)(C)CCCCCCCC.[Br-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1971

TSCA : No

SDS
Documents

Documents

Promotions

Promotions