missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyldioctylammonium Bromide 97.0+%, TCI America™
$288.94 - $723.25
Chemical Identifiers
| CAS | 3026-69-5 |
|---|---|
| Molecular Formula | C18H40BrN |
| Molecular Weight (g/mol) | 350.429 |
| MDL Number | MFCD00082931 |
| InChI Key | APTVNWGLSRAOFJ-UHFFFAOYSA-M |
| PubChem CID | 76408 |
| IUPAC Name | dimethyl(dioctyl)azanium;bromide |
| SMILES | CCCCCCCC[N+](C)(C)CCCCCCCC.[Br-] |
Chemical Identifiers
| 3026-69-5 | |
| 350.429 | |
| APTVNWGLSRAOFJ-UHFFFAOYSA-M | |
| dimethyl(dioctyl)azanium;bromide |
| C18H40BrN | |
| MFCD00082931 | |
| 76408 | |
| CCCCCCCC[N+](C)(C)CCCCCCCC.[Br-] |
Specifications
| 3026-69-5 | |
| C18H40BrN | |
| 5 g | |
| CCCCCCCC[N+](C)(C)CCCCCCCC.[Br-] | |
| 350.429 | |
| 350.43 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD00082931 | |
| APTVNWGLSRAOFJ-UHFFFAOYSA-M | |
| dimethyl(dioctyl)azanium;bromide | |
| 76408 | |
| ≥97.0% (T) | |
| Dimethyldioctylammonium Bromide |
Safety and Handling
EINECSNumber : (9)-1971
TSCA : No