missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylammonium Dimethyldithiocarbamate 95.0+%, TCI America™
$80.85 - $304.18
Chemical Identifiers
| CAS | 598-64-1 |
|---|---|
| Molecular Formula | C5H14N2S2 |
| Molecular Weight (g/mol) | 166.301 |
| MDL Number | MFCD00067267 |
| InChI Key | UVOFGKIRTCCNKG-UHFFFAOYSA-N |
| Synonym | Dimethyldithiocarbamic Acid Dimethylammonium Salt |
| PubChem CID | 11728 |
| IUPAC Name | dimethylazanium;N,N-dimethylcarbamodithioate |
| SMILES | C[NH2+]C.CN(C)C(=S)[S-] |
Chemical Identifiers
| 598-64-1 | |
| 166.301 | |
| UVOFGKIRTCCNKG-UHFFFAOYSA-N | |
| 11728 | |
| C[NH2+]C.CN(C)C(=S)[S-] |
| C5H14N2S2 | |
| MFCD00067267 | |
| Dimethyldithiocarbamic Acid Dimethylammonium Salt | |
| dimethylazanium;N,N-dimethylcarbamodithioate |
Specifications
| 598-64-1 | |
| White-Yellow | |
| MFCD00067267 | |
| Dimethyldithiocarbamic Acid Dimethylammonium Salt | |
| C[NH2+]C.CN(C)C(=S)[S-] | |
| 166.301 | |
| 166.30 | |
| Crystalline Powder |
| 132°C | |
| C5H14N2S2 | |
| 5 g | |
| UVOFGKIRTCCNKG-UHFFFAOYSA-N | |
| dimethylazanium;N,N-dimethylcarbamodithioate | |
| 11728 | |
| ≥95.0% (T) | |
| Dimethylammonium Dimethyldithiocarbamate |
Safety and Handling
EINECSNumber : (2)-2893
RTECSNumber : FA0250000
TSCA : Yes