accessibility menu, dialog, popup

UserName

H3CNHCH3H3CNCH3SHS

CAS RN 598-64-1

IUPAC Name: dimethyl[sulfanyl(carbonothioyl)]amine; dimethylamine
Synonyms: dimethyl[sulfanyl(carbonothioyl)]amine; dimethylamine
Molecular Weight (g/mol): 166.3
Molecular Formula: C5H14N2S2
InChi Key: UVOFGKIRTCCNKG-UHFFFAOYSA-N
SMILES: CNC.CN(C)C(S)=S
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results