missing translation for 'onlineSavingsMsg'
Learn More

Dimethylammonium Dimethyldithiocarbamate 95.0+%, TCI America™

Catalog No. D07155G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D07155G 5 g
D071525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D07155G Supplier TCI America Supplier No. D07155G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 598-64-1
Molecular Formula C5H14N2S2
Molecular Weight (g/mol) 166.301
MDL Number MFCD00067267
InChI Key UVOFGKIRTCCNKG-UHFFFAOYSA-N
Synonym Dimethyldithiocarbamic Acid Dimethylammonium Salt
PubChem CID 11728
IUPAC Name dimethylazanium;N,N-dimethylcarbamodithioate
SMILES C[NH2+]C.CN(C)C(=S)[S-]

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 5 g
Formula Weight 166.30
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylammonium Dimethyldithiocarbamate
EINECSNumber (2)-2893
RTECSNumber FA0250000
TSCA Yes