missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
| Color | Colorless to Yellow |
| Density | 1.0500g/mL |
| Boiling Point | 60°C to 61°C |
| Flash Point | 13°C |
| Infrared Spectrum | Authentic |
| Packaging | Glass bottle |
| Refractive Index | 1.4520 to 1.456 |
| Linear Formula | ((CH3)2NH2)·(CH3)2NCO2) |
| Quantity | 500 g |
| Specific Gravity | 1.05 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if inhaled. Harmful if swallowed. Causes severe skin burns and eye damage. Highly flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsi |
RUO – Research Use Only