accessibility menu, dialog, popup

UserName

H3CNHCH3H3CNCH3OHO

CAS RN 4137-10-4

IUPAC Name: dimethylamine; dimethylcarbamic acid
Synonyms: carbamic acid, dimethyl-; dimethylamine
Molecular Weight (g/mol): 134.18
Molecular Formula: C5H14N2O2
InChi Key: JIYXHCMRGZVYMA-UHFFFAOYSA-N
SMILES: CNC.CN(C)C(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)