missing translation for 'onlineSavingsMsg'
Learn More

Dimethylammonium-dimethylcarbamate, ratio carbon dioxide/dimethylamine <2:1

Catalog No. AC292901000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC292901000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292901000 Supplier Thermo Scientific Chemicals Supplier No. 292901000
Only null left

CAS: 4137-10-4 | C5H14N2O2 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Chemical Identifiers

CAS 4137-10-4
Molecular Formula C5H14N2O2
Molecular Weight (g/mol) 134.18
MDL Number MFCD00029888
InChI Key JIYXHCMRGZVYMA-UHFFFAOYSA-N
Synonym DIMCARB
IUPAC Name dimethylamine; dimethylcarbamic acid
SMILES CNC.CN(C)C(O)=O

Specifications

Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 60°C to 61°C
Flash Point 13°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4520 to 1.456
Linear Formula ((CH3)2NH2)·(CH3)2NCO2)
Quantity 100 g
Specific Gravity 1.05
Solubility Information Solubility in water: may decompose. Other solubilities: forming carbon dioxide,dimethylamine
Viscosity 84.3 mPa.s (20°C)
Formula Weight 134.18
Physical Form Liquid
Chemical Name or Material Dimethylammonium-dimethylcarbamate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only