missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaminophosphoryl Dichloride, TCI America™

Supplier:  TCI America P09535G

 View more versions of this product

Catalog No. P09535G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dimethylaminophosphoryl Dichloride
Yellow
C2H6Cl2NOP
5 g
dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride
CN(C)P(=O)(Cl)Cl
161.95
161.95
Liquid
677-43-0
195°C
MFCD00013896
3265
YNHXBEVSSILHPI-UHFFFAOYSA-N
N-dichlorophosphoryl-N-methylmethanamine
12673
≥98.0% (GC,T)

Chemical Identifiers

677-43-0
161.95
YNHXBEVSSILHPI-UHFFFAOYSA-N
12673
CN(C)P(=O)(Cl)Cl
C2H6Cl2NOP
MFCD00013896
dimethylphosphoramidic dichloride, n,n-dimethylphosphoramidodichloridate, dichlorophosphoric dimethylamide, phosphoramidic dichloride, dimethyl, dimethylamido phosphoric dichloride, dimethylamidophosphoric acid dichloride, dimethylaminophosphoryl dichloride, n,n-dimethylphosphoramidic dichloride, dimethylamidophosphoric dichloride, n,n-dimethylphosphoramic dichloride
N-dichlorophosphoryl-N-methylmethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TB4025000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions