missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine (ca. 11% in Ethanol, ca. 2.0mol/L), TCI America™

Catalog No. D3936500ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3936500ML 500 mL
D3936100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3936500ML Supplier TCI America Supplier No. D3936500ML

Chemical Identifiers

CAS 124-40-3
Molecular Formula C2H7N
Molecular Weight (g/mol) 45.085
MDL Number MFCD00008288
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
PubChem CID 674
ChEBI CHEBI:17170
IUPAC Name N-methylmethanamine
SMILES CNC

Specifications

Color Colorless
Quantity 500 mL
UN Number 2924
Formula Weight 45.09
Physical Form Liquid
Chemical Name or Material Dimethylamine (ca. 11% in Ethanol, ca. 2.0mol/L)
EINECSNumber (2)-0134
RTECSNumber IP8750000
TSCA Yes
Recommended Storage Refrigerator