missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine, 2M in THF, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethylamine is used as a precursor in the preparation of nitrosodimethylamine.
Solubility
Miscible with water and most organic solvents.
Notes
Hygroscopic. Incompatible with oxidizing agents and oxygen,Sodium aluminum hydride, sodium tetrahydroaluminate, lithium tetrahydroaluminate, borane, metal halides, strong acids, caustic alkalis and bromine.
Dimethylamine is used as a precursor in the preparation of nitrosodimethylamine.
Solubility
Miscible with water and most organic solvents.
Notes
Hygroscopic. Incompatible with oxidizing agents and oxygen,Sodium aluminum hydride, sodium tetrahydroaluminate, lithium tetrahydroaluminate, borane, metal halides, strong acids, caustic alkalis and bromine.
Specifications
Specifications
| Density | 0.85 |
| Boiling Point | ∼60°C |
| Flash Point | −36°C (−33°F) |
| Linear Formula | (CH3)2NH |
| Quantity | 100 mL |
| UN Number | UN2924 |
| Beilstein | 605257 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,3228 |
| Solubility Information | Miscible with water and most organic solvents. |
| Show More |
Safety and Handling
| Hazard Category | H225-H302-H314-H335-H336-H351 |
| Hazard Statement | GHS H Statement H225-H314-H351-H335 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. |
| Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only