missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 2M in THF, Thermo Scientific Chemicals

Catalog No. AAH27665AU
Change view
Click to view available options
Quantity:
100 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27665AU 1000 mL
AAH27665AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27665AU Supplier Thermo Scientific Chemicals Supplier No. H27665AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylamine is used as a precursor in the preparation of nitrosodimethylamine.

Solubility
Miscible with water and most organic solvents.

Notes
Hygroscopic. Incompatible with oxidizing agents and oxygen,Sodium aluminum hydride, sodium tetrahydroaluminate, lithium tetrahydroaluminate, borane, metal halides, strong acids, caustic alkalis and bromine.

Chemical Identifiers

CAS 124-40-3
Molecular Formula C2H7N
Molecular Weight (g/mol) 45.085
MDL Number MFCD00008288
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
Synonym dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution
PubChem CID 674
ChEBI CHEBI:17170
IUPAC Name N-methylmethanamine
SMILES CNC

Specifications

Density 0.85
Boiling Point ∼60°C
Flash Point −36°C (−33°F)
Linear Formula (CH3)2NH
Quantity 1000 mL
UN Number UN2924
Beilstein 605257
Sensitivity Hygroscopic
Merck Index 14,3228
Solubility Information Miscible with water and most organic solvents.
Formula Weight 45.08
Concentration or Composition (by Analyte or Components) 2M in THF
Chemical Name or Material Dimethylamine
Show More Show Less
Hazard Category H225-H302-H314-H335-H336-H351
Hazard Statement GHS H Statement
H225-H314-H351-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only