missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethylamine, 26% w/v, Reagents
Dimethylamine, 26% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CD2611004C1
Specifications
| Dimethylamine, 26% w/v | |
| Colorless to Light Yellow | |
| Safety Coated Glass | |
| 4 L | |
| N-Methylmethanamine | |
| O | |
| 18.02 | |
| Reagent |
| 7732-18-5,124-40-3 | |
| 10 | |
| H2O | |
| 0.9 | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 26% | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| N-Methylmethanamine | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |