missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 10.4% (w/w) in Methanol, Reagents

Catalog No. 01336511
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336511 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336511 Supplier Labchem Inc Supplier No. C71959801C

Dimethylamine, 10.4% (w/w) in Methanol, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1 , 7732-18-5 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym N-Methylmethanamine
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless to Light Yellow
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 0.8149
Percent Purity 10.40%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Dimethylamine, 10.4% (w/w) in Methanol