missing translation for 'onlineSavingsMsg'
Learn More

γ,γ-Dimethylallyl pyrophosphate ammonium salt, ≥95.0% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111015260
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015260 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015260 Supplier Merck Emd Millipore Supplier No. 384265MG

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 69382-61-2
Physical Form Neat
Linear Formula C5H12O7P2 · xNH3
Synonym 3-Methyl-2-butenyl pyrophosphate ammonium salt; DMAPP; Prenyl pyrophosphate ammonium salt
Type Analytical Standard
Quantity 5 mg
Formula Weight 246.09 (free acid basis)
Percent Purity ≥95.0% (TLC)
Grade Analytical Standard