missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Yellow Indicator, Honeywell Fluka™
Indicator
$191.26 - $618.72
Chemical Identifiers
| CAS | 60-11-7 |
|---|---|
| Molecular Formula | C14H15N3 |
| Molecular Weight (g/mol) | 225.30 |
| MDL Number | MFCD00008308 |
| InChI Key | JCYPECIVGRXBMO-UHFFFAOYSA-N |
| PubChem CID | 6053 |
| ChEBI | CHEBI:17903 |
| IUPAC Name | N,N-dimethyl-4-phenyldiazenylaniline |
| SMILES | CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 60-11-7 | |
| 225.30 | |
| JCYPECIVGRXBMO-UHFFFAOYSA-N | |
| CHEBI:17903 | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 |
| C14H15N3 | |
| MFCD00008308 | |
| 6053 | |
| N,N-dimethyl-4-phenyldiazenylaniline |
Specifications
| 111°C | |
| C14H15N3 | |
| MFCD00008308 | |
| 746016 | |
| JCYPECIVGRXBMO-UHFFFAOYSA-N | |
| N,N-dimethyl-4-phenyldiazenylaniline | |
| 6053 | |
| 225.29g/mol | |
| Glass Bottle | |
| 5 to 7 (at 10.00 g/L 20°C) | |
| Dimethyl Yellow |
| 60-11-7 | |
| C6H5N=NC6H4N(CH3)2 | |
| UN2811 | |
| Insoluble in water | |
| CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1 | |
| 225.30 | |
| CHEBI:17903 | |
| Odourless | |
| Yellow | |
| 25 g |
Safety and Handling
P281-P301 + P310
H301-H351
Danger
DOTInformation : Class 6.1 - PG 3 - RQ - Toxic solids, organic, n.o.s. HI: all
EINECSNumber : 200-455-7
RTECSNumber : BX7350000