missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Tetrahydropyran-4,4-dicarboxylate 98.0+%, TCI America™

Supplier:  TCI America T182525G

 View more versions of this product

Catalog No. T182525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Dimethyl Tetrahydropyran-4,4-dicarboxylate
Colorless
MFCD02093499
Tetrahydropyran-4,4-dicarboxylic Acid Dimethyl Ester
COC(=O)C1(CCOCC1)C(=O)OC
202.206
202.21
Liquid
149777-00-4
C9H14O5
25 g
OTIZXJJKKRZQJR-UHFFFAOYSA-N
dimethyl oxane-4,4-dicarboxylate
11063483
≥98.0% (GC)

Chemical Identifiers

149777-00-4
202.206
OTIZXJJKKRZQJR-UHFFFAOYSA-N
11063483
COC(=O)C1(CCOCC1)C(=O)OC
C9H14O5
MFCD02093499
Tetrahydropyran-4,4-dicarboxylic Acid Dimethyl Ester
dimethyl oxane-4,4-dicarboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions