missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Tetrahydropyran-4,4-dicarboxylate 98.0+%, TCI America™

Catalog No. p-7136980
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18255G 5 g
T182525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18255G Supplier TCI America Supplier No. T18255G
Only null left

Chemical Identifiers

CAS 149777-00-4
Molecular Formula C9H14O5
Molecular Weight (g/mol) 202.206
MDL Number MFCD02093499
InChI Key OTIZXJJKKRZQJR-UHFFFAOYSA-N
Synonym Tetrahydropyran-4,4-dicarboxylic Acid Dimethyl Ester
PubChem CID 11063483
IUPAC Name dimethyl oxane-4,4-dicarboxylate
SMILES COC(=O)C1(CCOCC1)C(=O)OC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyl Tetrahydropyran-4,4-dicarboxylate