missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Tetradecanedioate 96.0+%, TCI America™
Supplier: TCI America T17265G
Specifications
| Dimethyl Tetradecanedioate | |
| 43°C | |
| 196°C | |
| MFCD00673459 | |
| tetradecanedioic acid, dimethyl ester, 1,14-dodecanedicarboxylic acid dimethyl ester, tetradecanedioic acid dimethyl ester, tetradecanedioic acid, 1,14-dimethyl ester, acmc-1cupl, 1,14-dimethyl tetradecanedioate, tetradecanedioic acid,1,14-dimethyl ester | |
| COC(=O)CCCCCCCCCCCCC(=O)OC | |
| 286.41 | |
| 286.41 | |
| Crystalline Powder |
| 5024-21-5 | |
| White | |
| C16H30O4 | |
| 5 g | |
| ZDJFDFNNEAPGOP-UHFFFAOYSA-N | |
| 1,14-dimethyl tetradecanedioate | |
| 78727 | |
| ≥96.0% (GC) |
Chemical Identifiers
| 5024-21-5 | |
| 286.41 | |
| ZDJFDFNNEAPGOP-UHFFFAOYSA-N | |
| 1,14-dimethyl tetradecanedioate |
| C16H30O4 | |
| MFCD00673459 | |
| 78727 | |
| COC(=O)CCCCCCCCCCCCC(=O)OC |
Safety and Handling
EINECSNumber : (2)-2630
TSCA : Yes