missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Terephthalate 99.0+%, TCI America™

Catalog No. T001525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T001525G 25 g
T0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T001525G Supplier TCI America Supplier No. T001525G

Chemical Identifiers

CAS 120-61-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008440
InChI Key WOZVHXUHUFLZGK-UHFFFAOYSA-N
Synonym dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate
PubChem CID 8441
IUPAC Name 1,4-dimethyl benzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Melting Point 142°C
Color White
Quantity 25 g
Formula Weight 194.19
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyl Terephthalate
EINECSNumber (3)-1328
RTECSNumber WZ1225000
TSCA Yes