missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide 99.0+%, TCI America™

Catalog No. p-7126437
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D079825G 25 g
D0798500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D079825G Supplier TCI America Supplier No. D079825G
Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Physical Form Liquid
Color Colorless
Quantity 25 g
Boiling Point 189°C
Formula Weight 78.13
Percent Purity ≥99.0% (GC)
Chemical Name or Material Dimethyl Sulfoxide
EINECSNumber (2)-1553
RTECSNumber PV6210000
TSCA Yes