missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, Puriss p.a, ACS Reagent, ≥99.9% (GC), Honeywell Riedel-de Haën™
Puriss. p.a., ACS Reagent,≥99.9% (GC)
$414.55 - $414.55
Chemical Identifiers
| CAS | 67-68-5 |
|---|---|
| Molecular Formula | C2H6OS |
| Molecular Weight (g/mol) | 78.13 |
| MDL Number | MFCD00002089 |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
| PubChem CID | 679 |
| ChEBI | CHEBI:28262 |
| SMILES | CS(C)=O |
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| CS(C)=O |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 |
Specifications
| 67-68-5 | |
| 1.10g/mL | |
| 42% | |
| Poly Bottle | |
| C2H6OS | |
| MFCD00002089 | |
| 0 to 0.1% (Coulometric) | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| ≥99.9% (GC) |
| Colorless | |
| 1 L | |
| 573°C | |
| 1.478 to 1.48 | |
| (CH3)2SO | |
| 0.42mmHg (20°C) | |
| 506008 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methylsulfinylmethane | |
| 679 | |
| 78.13g/mol | |
| Dimethyl sulfoxide |
Safety and Handling
RTECSNumber : 200-664-3