missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, MilliporeSigma™

Catalog No. 317275100ML
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
317275100ML 100 mL Glass Bottle
317275500ML 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 317275100ML Supplier MilliporeSigma Supplier No. 317275100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS Reagent Grade

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Density 1.10g/cm3 at 20°C
Quantity 100 mL
Flash Point 87°C (189°F)
Ignition Point 300°to302°C (572°to576°F)
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 0.08 kPa (0.6mm Hg) (at 20°C)
Boiling Point 189°C
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Percent Purity ≥99%
Grade ACS
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Recommended Storage Store in a segregated and approved area. Keep container in a cool, well-ventilated area. Keep container tightly closed and sealed until ready for use. Avoid all possible sourcesof ignition (spark or flame).