missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide 99.0+%, TCI America™
$53.90 - $127.07
Chemical Identifiers
| CAS | 67-68-5 |
|---|---|
| Molecular Formula | C2H6OS |
| Molecular Weight (g/mol) | 78.13 |
| MDL Number | MFCD00002089 |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
| PubChem CID | 679 |
| ChEBI | CHEBI:28262 |
| IUPAC Name | methanesulfinylmethane |
| SMILES | CS(C)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D5293100ML
|
TCI America
D5293100ML |
100 mL |
Each for $53.90
|
|
|||||
|
D5293500ML
|
TCI America
D5293500ML |
500 mL |
Each for $127.07
|
|
|||||
Chemical Identifiers
| 67-68-5 | |
| 78.13 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| 679 | |
| methanesulfinylmethane |
| C2H6OS | |
| MFCD00002089 | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CHEBI:28262 | |
| CS(C)=O |
Specifications
| 67-68-5 | |
| 189°C | |
| Clear Liquid at 20°C | |
| MFCD00002089 | |
| IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
| methanesulfinylmethane | |
| 679 | |
| 78.13 | |
| Dimethyl Sulfoxide [for Spectrophotometry] |
| Colorless | |
| 100 mL | |
| C2H6OS | |
| dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
| CS(C)=O | |
| 78.13 | |
| CHEBI:28262 | |
| ≥99.0% (GC) |
Safety and Handling
EINECSNumber : (2)-1553
RTECSNumber : PV6210000
TSCA : Yes