missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, For headspace gas chromatography SupraSolv™ MilliporeSigma™

Catalog No. M1019002500
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1019002500 2.5 L Glass Bottle
M1019000500 500 mL Glass Bottle
M1019001000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1019002500 Supplier Technidata Supplier No. 1.01900.2500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For headspace gas chromatography SupraSolv™

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Density 1.10g/cm3 at 20°C
Quantity 2.5 L
Flash Point 87°C (189°F)
Ignition Point 300°to302°C (572°to576°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.48
Vapor Pressure 0.6 hPa (20°C)
Boiling Point 189°C
Solubility Information Soluble in water: 1000g/L at 20°C
Formula Weight 78.13
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Recommended Storage 15° to 25°C (59° to 77°F); Tightly closed. Dry