missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Fisher BioReagents™

Catalog No. p-216495
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP231100 100 mL Amber Glass Bottle
BP2311 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP231100 Supplier Fisher BioReagents Supplier No. BPBP231100
Only null left

Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 % | CAS: 67-68-5 | C2H6OS | 78.13 g/mol

Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %

DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Absorbance ≤0.30 Absorbance Units at 280nm
Color Colorless
Decomposition Information >190°C
Density 1.100g/cm³
Quantity 100 mL
Identification Pass Test
Packaging Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99.7 %
Vapor Pressure 0.55mbar at 20°C
Linear Formula (CH3)2SO
Viscosity 1.98 mPa/s at 25°C
Water 0.2% max.
Merck Index 15, 3285
Boiling Point 189°C
Formula Weight 78.13
Percent Purity ≥99.7%
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0
Recommended Storage RT