missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice

Catalog No. 60046492
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046492 4 x 2.5 L
6002578 1 L
6002579 2.5 L
6002583 6 x 1 L
6002581 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.
Catalog No. 60046492 Supplier Solstice Supplier No. 348694X2.5L
Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Melting Point 16°C to 19°C (literature)
Absorbance max. 0.01 at 350 nm, max. 0.10 at 300 nm, max. 0.30 280 nm, max. 0.70 at 270 nm
CAS Max % 100.0000%
Density 1.10 g/mL(lit.)
Quantity 4 x 2.5 L
Flash Point 87°C
Packaging Glass Bottle
Linear Formula (CH3)2SO
Beilstein 506008
Boiling Point 189°C
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Percent Purity >99.7%
Grade HPLC
Chemical Name or Material Dimethyl sulfoxide, CHROMASOLV™ Plus, for HPLC, ≥99.7%
Show More Show Less
ShelfLife 1260 days from date of manufacture
EINECSNumber 200-664-3
Recommended Storage Room Temp