missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Solstice
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
Specifications
Specifications
| Melting Point | 16°C to 19°C |
| Density | 1.10g/mL |
| Quantity | 2.5 L |
| Refractive Index | n20/D 1.479 |
| Vapor Pressure | 0.42mmHg (20°C) |
| Linear Formula | (CH3)2SO |
| Beilstein | 506008 |
| Boiling Point | 189°C |
| IUPAC Name | methylsulfinylmethane |
| Formula Weight | 78.13g/mol |
| Show More |
Safety and Handling
| EINECSNumber | 200-664-3 |
| RTECSNumber | PV6210000 |