missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide (Certified ACS), Fisher Chemical™

Catalog No. D1281
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
HDPE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
D1281 1 L HDPE Bottle
D1284 4 L HDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1281 Supplier Thermo Fisher Scientific Supplier No. BPD1281
  • ONLINE EXCLUSIVE PRICE
    $378.91
    $121.87 / Each

    Save $257.04

    68.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • ONLINE EXCLUSIVE PRICE
    $2,273.46
    $731.22 / Case of 6 Each

    Save $1,542.24

    68.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Note: Sold for nonmedical use only. Not to be used or sold as a drug for humans or animals

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Acidity Pass Test (USP)
Appearance Clear, Colorless Liquid
Decomposition Information >190°C
Density 1.100g/cm³
Quantity 1 L
Flash Point 87°C (188.6°F)
Identification Pass Test (USP, EP/BP)
Packaging HDPE Bottle
Physical Form Liquid
Residue after Evaporation 0.01% max.
Assay Percent Range 99.9 % (CH3)2SO Minimum
Titratable Acid 0.001mEq/g max.
Vapor Pressure 0.55mbar at 20°C
Linear Formula (CH3)2SO
Viscosity 1.98 mPa/s at 25°C
Water 0.1% max.
Boiling Point 189°C
Solubility Information Soluble starch and sulfate: Pass Test (USP), 15 ppm max. (EP/BP)
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS
Concentration or Composition (by Analyte or Components) Dextrin: Pass Test (USP, EP/BP)
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0