missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ACS Reagent (DMSO), Reagents

Catalog No. p-200021163
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01338004 4 L Poly Bottle
01338003 4 L Glass Bottle
01338002 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01338004 Supplier Labchem Inc Supplier No. C11966504A
Only null left

Dimethyl Sulfoxide, ACS Reagent (DMSO), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym DMSO, Methylsulfinylmethane
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Boiling Point 189°C
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.101
Grade ACS
Chemical Name or Material Dimethyl Sulfoxide, ACS Reagent (DMSO)