missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, ≥99%, MP Biomedicals™

Catalog No. 361054181
Change view
Click to view available options
Quantity:
0.5 L
1 L
100 mL
25 mL
Catalog No. Quantity
361054181 1 L
361054178 25 mL
361054179 100 mL
361054180 0.5 L
4 options

Catalog No. 361054181

Supplier: MP Biomedicals Inc 0219605591

Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 1 L
Physical Form Liquid
Assay Percent Range ≥99%
IUPAC Name methanesulfinylmethane
Formula Weight 78.1
Percent Purity ≥99%
Chemical Name or Material Dimethyl sulfoxide