missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, ≥99%, MP Biomedicals™
Specifications
Specifications
| Quantity | 0.5 L |
| Physical Form | Liquid |
| Assay Percent Range | ≥99% |
| IUPAC Name | methylsulfinylmethane |
| Formula Weight | 78.1 |
| Percent Purity | ≥99% |
| Chemical Name or Material | Dimethyl sulfoxide |