missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, ≥99%, MP Biomedicals™

Catalog No. 361054178
Change view
Click to view available options
Quantity:
25 mL
100 mL
0.5 L
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361054178 25 mL
361054179 100 mL
361054180 0.5 L
361054181 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 361054178 Supplier MP Biomedicals Inc Supplier No. 0219605525
Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Quantity 25 mL
Physical Form Liquid
Assay Percent Range ≥99%
IUPAC Name methylsulfinylmethane
Formula Weight 78.1
Percent Purity ≥99%
Chemical Name or Material Dimethyl sulfoxide