missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, 99.97%, MP Biomedicals™

$80.21 - $272.07

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19481950
View Documents
MP Biomedicals Inc
0219481950
50 mL
Each for $80.21
Only null left
 
ICN19481983
View Documents
MP Biomedicals Inc
0219481983
250 mL
Each for $272.07
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CS(C)=O
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262

Specifications

67-68-5
1.10g/mL
Liquid To Solid
C2H6OS
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(C)=O
78.13
CHEBI:28262
Dimethyl sulfoxide
Colorless
50 mL
0.9997
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'