missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, 99.97%, MP Biomedicals™

Catalog No. ICN19481983
Change view
Click to view available options
Quantity:
50 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19481983 250 mL
ICN19481950 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19481983 Supplier MP Biomedicals Inc Supplier No. 0219481983
Only null left

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 250 mL
Physical Form Liquid To Solid
Assay Percent Range 0.9997
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Chemical Name or Material Dimethyl sulfoxide