missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ≥99.9%, Solstice

Catalog No. 60046948
Change view
Click to view available options
Quantity:
4 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60046948 4 L
6000537 6 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046948 Supplier Solstice Supplier No. 4723014L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS Reagent,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 4 L
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
RTECSNumber 200-664-3