Learn More
Dimethyl sulfate, 99+%, AcroSeal™
CAS: 77-78-1 | C2H6O4S | 126.13 g/mol
Supplier: Thermo Scientific Chemicals 430830010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
| Dimethyl sulfate | |
| -32.0°C | |
| 188.0°C | |
| 99+% | |
| C2H6O4S | |
| (CH3)2SO4 | |
| 1 L | |
| 15,328 | |
| Solubility in water: 2.8g/100mL (18°C). Other solubilities: hydrolysis rapidly at 18°C or above in water,soluble in ehter,dioxane,acetone and aromatic,hydrocarbons,sparingly soluble in carbon disulfide and,aliphatic hydrocarbons | |
| COS(=O)(=O)OC | |
| 126.13 | |
| 1.81 mPa.s (20°C) | |
| 126.13 |
| 77-78-1 | |
| 1.3300g/mL | |
| 83°C | |
| AcroSeal™ Glass Bottle | |
| 1.3860 to 1.3880 | |
| MFCD00008416 | |
| 1.33 | |
| dimethyl sulphate, sulfuric acid, dimethyl ester, dimethyl monosulfate, dimethylsulfate, dimethylsulfat, sulfate dimethylique, sulfuric acid dimethyl ester, dimethylsulfaat, dimetilsolfato, dms methyl sulfate | |
| VAYGXNSJCAHWJZ-UHFFFAOYSA-N | |
| dimethyl sulfate | |
| 6497 | |
| CHEBI:59050 | |
| 99+% |
Chemical Identifiers
| 77-78-1 | |
| 126.13 | |
| VAYGXNSJCAHWJZ-UHFFFAOYSA-N | |
| 6497 | |
| dimethyl sulfate |
| C2H6O4S | |
| MFCD00008416 | |
| dimethyl sulphate, sulfuric acid, dimethyl ester, dimethyl monosulfate, dimethylsulfate, dimethylsulfat, sulfate dimethylique, sulfuric acid dimethyl ester, dimethylsulfaat, dimetilsolfato, dms methyl sulfate | |
| CHEBI:59050 | |
| COS(=O)(=O)OC |
Safety and Handling
GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Fatal if inhaled.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory i
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
EINECSNumber : 201-058-1
RUO – Research Use Only