accessibility menu, dialog, popup

UserName

CH3OSOOOH3C

CAS RN 77-78-1

IUPAC Name: dimethyl sulfate
Synonyms: dimethyl sulfate sulfato de dimetilo dimethylsulfate DMS sulfate dimethylique
Molecular Weight (g/mol): 126.13
Molecular Formula: C2H6O4S
InChi Key: VAYGXNSJCAHWJZ-UHFFFAOYSA-N
SMILES: COS(=O)(=O)OC
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)