missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfate, 98%, Spectrum™ Chemical

Catalog No. p-7165460
Change view
Click to view available options
Quantity:
500 mL
1 L
200 L
Packaging:
Amber Glass Bottle
Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610215 500 mL Amber Glass Bottle
18610216 1 L Amber Glass Bottle
18610218 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18610215 Supplier Spectrum Chemical Mfg Cor Supplier No. D1045500MLGL
Only null left

Chemical Identifiers

CAS 77-78-1

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 0.98
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.