missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sebacate 98.0+%, TCI America™

Catalog No. p-7135771
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S002725G 25 g
S0027500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S002725G Supplier TCI America Supplier No. S002725G
Only null left

Chemical Identifiers

CAS 106-79-6
Molecular Formula C12H22O4
Molecular Weight (g/mol) 230.304
MDL Number MFCD00008472
InChI Key ALOUNLDAKADEEB-UHFFFAOYSA-N
Synonym dimethyl sebacate, methyl sebacate, decanedioic acid, dimethyl ester, sebacic acid dimethyl ester, sebacic acid, dimethyl ester, dimethyl octane-1,8-dicarboxylate, 1,10-dimethyl decanedioate, decanedioic acid dimethyl ester, decanedioic acid, 1,10-dimethyl ester, dimethyl decane-1,10-dioate
PubChem CID 7829
IUPAC Name dimethyl decanedioate
SMILES COC(=O)CCCCCCCCC(=O)OC

Specifications

Color White-Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 230.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Dimethyl Sebacate
EINECSNumber (2)-0879
TSCA Yes